Formaldehyde 37% in aqueous solution
Glass bottle, 1 L
N° de catalogue ROTH4979.1
Marketsource
Fournisseur: Roth Carl
Formaldehyde 37% in aqueous solution
N° de catalogue ROTH4979.1
Fournisseur: Roth Carl
Marketsource
Numéro CAS: 50-00-0
Jump to:
Product Details & Documents
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:2209
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



