Order Entry
Belgium
Orders LinkContactUsLinkComponent
1-Tetradecanol

1-Tetradecanol

Glass bottle, 1 L
Catalog # ROTH4741.2
Marketsource
Supplier:   Roth Carl
1-Tetradecanol
Catalog # ROTH4741.2
Supplier:  Roth Carl
Marketsource
CAS Number:  112-72-1
1-Tetradecanol

Glass bottle, 1 L

CAS Number:  112-72-1
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
  • CAS Number:112-72-1
  • Formula:H₃C(CH₂)₁₂CH₂OH
  • MDL Number:MFCD00004757
  • Molecular Weight:214.39 g/mol
  • Merck Index:13,06361
  • Boiling Pt:289 °C (1013 hPa)
  • Melting Pt:37…39 °C
  • Density:0.8 g/cm³ (60 °C)
  • Storage Temperature:Ambient
  • EINECS:204-000-3
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.