L(+)-Glutamine
Glass bottle for solids, 500 g
Catalog # ROTH3772.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
- CAS Number:56-85-9
- Formula:C₅H₁₀N₂O₃
- MDL Number:MFCD00008044
- Molecular Weight:146.15 g/mol
- Merck Index:13,04484
- Boiling Pt:355 °C (1013 hPa)
- Melting Pt:185…186 °C
- Density:1.5326 g/cm³ (130 °C)
- Storage Temperature:Ambient
- EINECS:200-292-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



