Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

L(+)-Glutamine

Glass bottle for solids, 100 g
Catalog # ROTH3772.1
Marketsource
Supplier:   Roth Carl
L(+)-Glutamine
Catalog # ROTH3772.1
Supplier:  Roth Carl
Marketsource
CAS Number:  56-85-9
L(+)-Glutamine
L(+)-Glutamine

Glass bottle for solids, 100 g

CAS Number:  56-85-9
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
  • CAS Number:56-85-9
  • Formula:C₅H₁₀N₂O₃
  • MDL Number:MFCD00008044
  • Molecular Weight:146.15 g/mol
  • Merck Index:13,04484
  • Boiling Pt:355 °C (1013 hPa)
  • Melting Pt:185…186 °C
  • Density:1.5326 g/cm³ (130 °C)
  • Storage Temperature:Ambient
  • EINECS:200-292-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications