Hydroquinone
Plastic bottle for solids, 100 g
Catalog # ROTH3586.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 1,4-Dihydroxybenzene, Quinol
- CAS Number:123-31-9
- Formula:C₆H₄(OH)₂
- MDL Number:MFCD00002339
- Molecular Weight:110.11 g/mol
- Merck Index:13,04833
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:172 °C
- Density:1.332 g/cm³ (20 °C)
- Flash Pt:165 °C
- Storage Temperature:Ambient
- EINECS:204-617-8
- UN:3077
- ADR:9,III
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g



