Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid

L(+)-Ascorbic acid

Glass bottle for solids, 250 g
N° de catalogue ROTH3525.2
Marketsource
Fournisseur:   Roth Carl
L(+)-Ascorbic acid
N° de catalogue ROTH3525.2
Fournisseur:  Roth Carl
Marketsource
Numéro CAS:  50-81-7
L(+)-Ascorbic acid
L(+)-Ascorbic acid

Glass bottle for solids, 250 g

Numéro CAS:  50-81-7
Prix
21,90 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…192 °C
  • Density:1.694 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.