Agar
Glass bottle for solids, 1 kg
Catalog # ROTH2266.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:13,00184
- Melting Pt:90 °C
- Storage Temperature:Ambient
- EINECS:232-658-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



