Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
10 ml, 0,05 ml, 0,030 ± ml
Catalog # BURK9695-1010
Supplier: BURKLE
Automatic burettes, Schilling pattern, Schellbach stripe, LaboPlast®
Catalog # BURK9695-1010
Supplier: BURKLE
Aller à :
Product Details & Documents
Borosilicate glass 3.3, class B.
- Adjusted for Ex
- Blue ring graduations, Schellbach stripe
- Precise dosing using the press button, exact fine titration with the micro-screw and precise discharge jet
Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Certifications: Graduation according to Category B (Tolerances according to DIN EN ISO 385).
Documents
Spécifications
- Capacity:10 ml
- Division:0,05 ml
- Tolerance:0,030 ± ml
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Spare nozzle, PP
Informations sur le produit
- Catalog #AvailabilityPrice
- Cap without feed tube for 1000 ml bottleCap without feed tube for 1000 ml bottleBURK9695-0528Connexion Pour la disponibilité1 * 1 ST - 2,40 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



