Orders LinkContactUsLinkComponent
Biochemical ID test systems
Biochemical ID test systems
Identification System Software (version 1.0.11.13) for use with both ID Test Systems (PC only, Windows 95 or later), 1
Catalog # 709-0258
Supplier:   TECRA
Biochemical ID test systems
Catalog # 709-0258
Supplier:  TECRA
CAS Number:  
Biochemical ID test systems
Biochemical ID test systems
Identification System Software (version 1.0.11.13) for use with both ID Test Systems (PC only, Windows 95 or later), 1
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
ID kits can be used to identify pure cultures but are also useful for the identification and confirmation of isolates that have produced presumptive positive results in any of TECRA’s Salmonella or Listeria rapid screening kits. The ID kits employ just 12 standardised microwell substrates. They are simple to use, with no assembly required, single colony inoculation and non-toxic reagents. The Identification System Software can be used to provide a reliable identification.
  • The kits are conveniently sized and contain all the reagents required
  • ID kits can save time and money, eliminating the need to make up numerous types of biochemical test media
  • Greater accuracy and reliability
Specifications
Specifications
  • Pk:
    1
  • Description:
    Identification System Software (version 1.0.11.13) for use with both ID Test Systems (PC only, Windows 95 or later)
Specifications