TEST TUBE 10MLCYLINDRICAL PP 16X100MM 1 * 2.000 items
Catalog # LPIT111040
Supplier: LP ITALIANA
Frequently Bought Together
Sulphuric acid 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
Supplier: VWR Collection
Finned plugs for tubes
Supplier: KARTELL
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Magnetic stirring bars, cylindrical
Supplier: VWR Collection
Devarda's alloy, granules, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Repetitive pipette, Multipette® M4
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Bottle top vacuum filtration systems, Nalgene®
Supplier: Thermo Fisher Scientific
Syringeless filters, Mini-UniPrep™, Whatman™
Supplier: Whatman products (Cytiva)