Order Entry
Belgium
Orders LinkContactUsLinkComponent
Transfer pipettes, fine tip

Transfer pipettes, fine tip

Fine tip, maxi, 4,8 ml, 22 µl, 147 mm, +, Bulk
  ALPCLW4236
Transfer pipettes, fine tip
  ALPCLW4236
Transfer pipettes, fine tip
Transfer pipettes, fine tip

Fine tip, maxi, 4,8 ml, 22 µl, 147 mm, +, Bulk

 
171,00 €
   
1 * 3.200 ST
 

Non toxic, food grade LDPE.

  • For careful control and small volume transfers
  • Ideal for supernatant removal, slide preparation and gel loading
 
  • Packed:
    Bulk
  • Sterile:
    +
  • Capacity:
    4,8 ml
  • Description:
    Fine tip, maxi
  • Length:
    147 mm
  • Seq:
    13
  • Drop vol.:
    22 µl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.