Orders LinkContactUsLinkComponent
Complement reagents
Rat complement
Catalog # ROCKC205-0005
Complement reagents
Catalog # ROCKC205-0005
Supplier:  
Complement reagents
Complement reagents
Rat complement
Discontinued
Product Details & Documents
Complement reagents are prepared from healthy, non-medicated animals using very large pools to reduce lot-to-lot variability. The fresh frozen complement is rapidly processed and maintained at –70 °C to ensure maximum activity. The lyophilised complement is supplied with a diluent of veronal buffered saline containing optimal concentrations of magnesium and calcium.
Specifications
  • Description:
    Rat complement
  • Seq:
    27
  • Cat. No.:
    ROCKC205-0005
  • Formulation:
    Lyophilized