Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flat optical windows

Flat optical windows

Flat optics disc, Quartz (Suprasil® 300), 22 mm, 1,25 mm
Catalog # HELL202-46
Supplier:   Hellma
Flat optical windows
Catalog # HELL202-46
Supplier:  Hellma
Flat optical windows
Flat optical windows

Flat optics disc, Quartz (Suprasil® 300), 22 mm, 1,25 mm

Price
64,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Optical glass cells and elements dedicated to special applications with detachable window.

Suitable for high precision analytical techniques.

Specifications
  • Window material:
    Quartz (Suprasil® 300)
  • Description:
    Flat optics disc
  • Dimensions:
    22 mm
  • Thickness:
    1,25 mm