Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Standard opening screw thread vials

Vials, without writing patch, 2,00 ml, Flat bottom, 12×32 mm, Glass, Clear, No, 8-425
  NSCAC4013-1
Standard opening screw thread vials
  NSCAC4013-1
Standard opening screw thread vials
Standard opening screw thread vials

Vials, without writing patch, 2,00 ml, Flat bottom, 12×32 mm, Glass, Clear, No, 8-425

 
25,30 €
   
1 * 100 ST
 

Preferred for use with many carousel tray autosamplers. Glass vials are manufactured from superior quality borosilicate clear glass Type 1.

  • Re-usable sleeves

Microsampling and high recovery vials allow maximum sample extraction without need for separate inserts.

 
  • Cap included:
    No
  • Cap Size:
    8-425
  • Ø×H:
    12×32 mm
  • Capacity:
    2,00 ml
  • Colour:
    Clear
  • Description:
    Vials, without writing patch
  • Material:
    Glass
  • Seq:
    02
  • Base:
    Flat bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.