Order Entry
Belgium
Orders LinkContactUsLinkComponent
Retord rod, without thread

Retord rod, without thread

13 mm, 610 mm
Catalog # 241-7870
Supplier:   R L ENTERPRISES
Retord rod, without thread
Catalog # 241-7870
Supplier:  R L ENTERPRISES
Retord rod, without thread
Retord rod, without thread

13 mm, 610 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Anodized aluminium.

Ø = 13 mm

Specifications
  • Ø:
    13 mm
  • Length:
    610 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.