Orders LinkContactUsLinkComponent
Watchmakers' magnifiers
Watchmakers' magnifiers
Magnifier with dual lens, 13 mm
Catalog # ESCH1130
Supplier:   ESCHENBACH OPTIK
Watchmakers' magnifiers
Catalog # ESCH1130
Supplier:  ESCHENBACH OPTIK
Watchmakers' magnifiers
Watchmakers' magnifiers
Magnifier with dual lens, 13 mm
Price
48,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
PXM® lightweight lenses with antistatic and scratch resistant coating. Ideal as a watchmaker's magnifier or gemstone magnifier.
  • Includes vent to prevent condensation
  • Made of flame resistant plastic material
  • Colour: anthracite
Specifications
  • Ø:
    13 mm
  • Magnification:
    15×
  • Description:
    Magnifier with dual lens
  • Seq:
    004