Orders LinkContactUsLinkComponent
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
1000 ml, 29/32 NS, 14/23 NS
Catalog # WITG0.768.001
Supplier:   witeg Labortechnik
Flasks, round bottom, with three parallel necks
Catalog # WITG0.768.001
Supplier:  witeg Labortechnik
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
1000 ml, 29/32 NS, 14/23 NS
Price
69,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Specifications
  • Capacity:
    1000 ml
  • Side necks:
    14/23 NS
  • Centre neck:
    29/32 NS