Order Entry
Belgium
Orders LinkContactUsLinkComponent
Funnels, micro

Funnels, micro

8 mm, 35 mm
Catalog # 221-1250
Funnels, micro
Catalog # 221-1250
Funnels, micro
Funnels, micro

8 mm, 35 mm

Price
64,20 €
Log in for availability
1 * 12 ST
Product Details & Documents

PP

  • Good chemical resistance
  • Autoclavable at 121 °C

Ideal for filling burettes and small bottles.  

Specifications
  • Funnel O-Ø:
    35 mm
  • Stem O-Ø:
    8 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.