Order Entry
Belgium
Orders LinkContactUsLinkComponent
Starch (from corn)

Starch (from corn)

5 kg
Catalog # ROTH9444.2
Marketsource
Supplier:   Roth Carl
Starch (from corn)
Catalog # ROTH9444.2
Supplier:  Roth Carl
Marketsource
CAS Number:  9005-25-8
Starch (from corn)

5 kg

CAS Number:  9005-25-8
Price
Inquire For Price
Log in for availability
1 * 5 kg
Product Details & Documents
  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-679-6

Documents

Specifications

Specifications

  • Pk:
    5 kg

Specifications