Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrodes kit, 2-Needle Array™

Electrodes kit, 2-Needle Array™

Two needle array kit, 5 mm gap
Catalog # 732-0861
Supplier:   BTX
Electrodes kit, 2-Needle Array™
Catalog # 732-0861
Supplier:  BTX
Electrodes kit, 2-Needle Array™
Electrodes kit, 2-Needle Array™

Two needle array kit, 5 mm gap

Price
624,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These in vivo style electrode specifically designed for muscle gene delivery. Among the non-viral techniques for in vivo gene transfer, the direct injection of plasmid DNA into muscle is simple, inexpensive and safe. The electrode is available in two gap sizes, 5 mm and 10 mm.

  • Handle made of Delrin, needle made of stainless steel
  • 5 mm 2-Needle array and handle is designed for small muscle masses such as mouse tibialis
  • 10 mm 2-Needle array and handle is designed for larger muscle masses such as rat gastrocnemius

Generator compatibility: ECM 830 and ECM 2001

Voltage range: 0 to 500 V

Pulse length range: 10 µsec to 90 msec

Needle length: 20 mm

Handle length: 8 cm

Delivery information: Electrodes are supplied in a convenient kit that includes one 2-Needle Array handle and six 2-Needle Arrays

Specifications
  • Application:
    In vivo drug or gene delivery, muscle gene therapy
  • Description:
    Two needle array kit, 5 mm gap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.