Conical separating funnels
50 ml, 14/23 NS
Catalogus # 532-0103
Leverancier: SciLabware
Ga naar:
Product Details & Documents
PYREX®, borosilicate glass, with glass stopper.
With ground glass stopper and interchangeable PTFE key. Without graduation.
Specificaties
- Socket size:14/23 NS
- Capacity:50 ml
- Seq:01