Roller bottles, Nunc™ InVitro™
Expanded surface (2.5X), 2100 cm², 200-600 ml
Catalog # 734-2397
Supplier: Thermo Fisher Scientific
Roller bottles, Nunc™ InVitro™
Catalog # 734-2397
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PETG, sterile.
- Quick-action ergonomic closure reduces wrist stain and increases productivity
- Wide range of sizes with surface areas from 1050 to 4200 cm²
- Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
- Easy-to-read graduations for medium fills
- Lot number is printed on each bottle to maximise traceability
For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Specifications
- Growth area:2100 cm²
- Recommended working volume:200-600 ml
- Description:Expanded surface (2.5X)
- Cat. No.:734-2397
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact



