Roller bottles, Nunc™ InVitro™
Expanded surface (2X), 1700 cm², 200-600 ml
Catalog # 734-2396
Supplier: Thermo Fisher Scientific
Roller bottles, Nunc™ InVitro™
Catalog # 734-2396
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
PETG, sterile.
- Quick-action ergonomic closure reduces wrist stain and increases productivity
- Wide range of sizes with surface areas from 1050 to 4200 cm²
- Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
- Easy-to-read graduations for medium fills
- Lot number is printed on each bottle to maximise traceability
For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Documents
Specifications
- Growth area:1700 cm²
- Recommended working volume:200-600 ml
- Description:Expanded surface (2X)
- Cat. No.:734-2396
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



