Order Entry
Belgium
Orders LinkContactUsLinkComponent
Roller bottles, Nunc™ InVitro™

Roller bottles, Nunc™ InVitro™

Standard PDL-coated, vented (1.2X), 1050 cm², 100-500 ml
Catalog # 734-2394
   Low Manufacturing Impact
Roller bottles, Nunc™ InVitro™
Catalog # 734-2394
   Low Manufacturing Impact
Roller bottles, Nunc™ InVitro™
Roller bottles, Nunc™ InVitro™

Standard PDL-coated, vented (1.2X), 1050 cm², 100-500 ml

Price
211,00 €
Log in for availability
1 * 20 ST
Product Details & Documents

PETG, sterile.

  • Quick-action ergonomic closure reduces wrist stain and increases productivity
  • Wide range of sizes with surface areas from 1050 to 4200 cm²
  • Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
  • Easy-to-read graduations for medium fills
  • Lot number is printed on each bottle to maximise traceability

For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.

Specifications
  • Growth area:
    1050 cm²
  • Recommended working volume:
    100-500 ml
  • Description:
    Standard PDL-coated, vented (1.2X)
  • Cat. No.:
    734-2394
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact