Roller bottles, Nunc™ InVitro™
Standard (1.2X), 1050 cm², 100-500 ml
N° de catalogue 734-2393
Fournisseur: Thermo Fisher Scientific
Roller bottles, Nunc™ InVitro™
N° de catalogue 734-2393
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
PETG, sterile.
- Quick-action ergonomic closure reduces wrist stain and increases productivity
- Wide range of sizes with surface areas from 1050 to 4200 cm²
- Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
- Easy-to-read graduations for medium fills
- Lot number is printed on each bottle to maximise traceability
For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.
Attention: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Documents
Spécifications
- Growth area:1050 cm²
- Recommended working volume:100-500 ml
- Description:Standard (1.2X)
- Cat. No.:734-2393
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact



