Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
pH Papers, without color scale
: MACHEREY-NAGEL
Buffer solutions in sachets
: Mettler - Toledo
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, ready to use
: WTW