Orders LinkContactUsLinkComponent
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
Orange, Cellophane wrapped slides
Catalogus # 630-2100
Leverancier:   Epredia
Microscope slides, SuperFrost®, Menzel Gläser
Catalogus # 630-2100
Leverancier:  Epredia
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
Orange, Cellophane wrapped slides
Price
8,40 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
  • Labelling area resistant to all common solvents
  • Six colours for identification and colour coding
  • Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Certificeringen: ISO 8037/I.
Verpakking: In cardboard boxes of 50.
Specificaties
  • Packed:
    Cellophane wrapped slides
  • Colour:
    Orange
  • Seq:
    319