Orders LinkContactUsLinkComponent
Bellow connectors
Bellow connectors
34/35 NS, 34/35 NS
Catalog # KART1049
Supplier:   KARTELL
Bellow connectors
Catalog # KART1049
Supplier:  KARTELL
Bellow connectors
Bellow connectors
34/35 NS, 34/35 NS
Price
302,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Bellow connector, PTFE, for use as a flexible or elastic connector to prevent pressure and stresses, and to dampen vibrations in standard ground joints.
  • Inert
  • Temperature resistant up to 280 °C
  • Very good chemical resistance
With ground joints on cone and socket. Sturdy construction with trapezoidal sealing rings, which give medium to high vacuum performance.
Specifications
  • Cone size:
    34/35 NS
  • Socket size:
    34/35 NS
  • Seq:
    03