Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH/°C testers, 206

pH/°C tester, 206 pH3, single instrument
Catalog # TEST0563.2063
Supplier:   TESTO
pH/°C testers, 206
Catalog # TEST0563.2063
Supplier:  TESTO
pH/°C testers, 206
pH/°C testers, 206

pH/°C tester, 206 pH3, single instrument

Price
127,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The testo 206 series is ideal for pH measurements in liquid and semi-solid media. The instruments offer a combination of a pH immersion probe and a temperature probe for precise and fast temperature compensation. Thanks to the large volume of gel electrolyte and the dual wall diaphragm, the probe is leak-proof, maintenance-free and insensitive to dirt.

  • Built-in temperature sensor
  • Maintenance-free gel electrolyte
  • Robust, waterproof and dishwasher-safe protective cover (TopSafe, protection class IP 65)
  • Automatic final value recognition (Auto-hold)
  • 1, 2, or 3-point calibration possible

Testo 206-pH1 is specially designed for fast pH checks in liquids.

Testo 206-pH2 is ideally suited for pH measurement in semi-solid and protein-rich foods such as jellies, creams, cheese, fruit etc.

Testo 206-pH3 is equipped with a BNC socket which allows the connection of any pH probes to the instrument, depending on the application. This makes the instrument universally applicable.

 : Single instruments are supplied with TopSafe cover and belt/wall holder. pH1 and pH2 also include a storage cap with gel.
pH1 and pH2 starter kits are supplied with storage cap with gel, calibration dosing bottles 250 ml pH 4 and 7, TopSafe cover, belt/wall holder and aluminium case.
All instruments are supplied with probe head but without probe. Probes need to be ordered separately.

Specifications
  • Description:
    pH/°C tester, 206 pH3, single instrument
  • Compliance:
    IP68
  • pH range:
    0...+14
  • Power supply:
    1 × CR2032 battery
  • Temperature compensation:
    Automatic
  • pH calibration:
    1, 2, or 3-point calibration
  • Temperature resolution:
    0,1 °C
  • Temperature accuracy:
    ±0,4 °C
  • Weight:
    0,062 kg
  • Cat. No.:
    TEST0563.2063
  • pH accuracy:
    ±0,02
  • pH resolution:
    0,01
  • Model:
    206 pH3
  • W×D×H:
    33×20×197 mm
  • Temperature range:
    Depending on sensor
  • Display:
    LCD, 2 lines
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Storage cap for testo 206, with KCI gel filling
    Storage cap for testo 206, with KCI gel filling
    TEST0554.2067
    Log in for availability
    1 * 1 ST - 17,00 €

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer solution 7,00 in dosing bottle (250 ml), with DAkkS calibration certificate
    pH buffer solution 7,00 in dosing bottle (250 ml), with DAkkS calibration certificate
    TEST0554.2063
    Log in for availability
    1 * 250 mL - 21,00 €

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer solution 4,01 in dosing bottle (250 ml), with DAkkS calibration certificate
    pH buffer solution 4,01 in dosing bottle (250 ml), with DAkkS calibration certificate
    TEST0554.2061
    Log in for availability
    1 * 250 mL - 21,00 €