VWR® PureStep, Multi-Layer Adhesive Floor Mats
Product Details & Documents
30 or 60 layer adhesive entrance mats manufactured from low density polyethylene. These mats control contamination by removing debris from the soles of shoes, and the wheels of transport containers. When one layer is dirty, simply remove by the numbered corner tab, leaving a fresh layer ready for use.
- Antimicrobial agent protects against the growth of bacteria and fungi
- Non slip, low profile, individually numbered layers
- Reduced maintenance and cleaning costs
- Contains GMO-free, renewable plant-based material that reduces CO₂ emissions
- Adhesive mats are also recyclable after use
Mats are available in 30 layer, 60 layer, 30 layer high tack, and 60 layer high tack options. Colour: Clear, white, grey or blue.
Used mat layers, including top and bottom release liners, can be recycled after use using the Recycling solution for plastic packaging, Zero Waste Box™ from TerraCycle.
Documents
Antimicrobial report on PureStep multilayer adhesive mats Microbial adherence report for adhesive mats What is the environmental impact of your business?
Avantor Purestep Adhesive Mats
Documentation produit
CoC Multi-layer Adhesive Floor Mats, PureStep 1.8 mm Thickness.pdf CoC VWR Multi-Layer Adhesive Floor Mats PureStep.pdf
Déclaration de conformité
CoC VWR Multi-Layer Adhesive Floor Mats, PureStep COC_VWRI115-0047_UR0317 COC_VWRI115-0047_UU0249 coc_VWRI115-0049_UT0261 coc_VWRI115-0049_UT0304
VWRI115-0057_TDS_EN VWRI115-0063_TDS_EN VWRI115-0072_TDS_EN
Fiche technique
Specifications
- Colour:Blue
- L×W:610×762 mm
- Seq:09
- Thickness:8 µm ±20%
Accessories
Product Information
- Catalog #ColourForAvailabilityPrice
- Mat frame
Product Information
- Catalog #ColourForAvailabilityPrice
- Mat frame
Product Information
- Catalog #ColourForAvailabilityPrice
- Mat frame
Product Information
- Catalog #ColourForAvailabilityPrice
- Mat frame
Product Information
- Catalog #ColourForAvailabilityPrice
- Mat frame
Product Information
- Catalog #ColourForAvailabilityPrice
- Mat frame
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



