Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, rectangular

VWR®, Magnetic stirring bars, rectangular

25 mm, 5,5 mm
Catalog # 442-0487
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, rectangular
Catalog # 442-0487
Supplier:  VWR Collection
VWR®, Magnetic stirring bars, rectangular
VWR®, Magnetic stirring bars, rectangular

25 mm, 5,5 mm

Price
28,30 €
Log in for availability
1 * 10 ST
Product Details & Documents

PTFE covered, strong Alnico V magnetic core. Smooth surface. Very effective stirrers, especially at low speeds.

  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Specifications
  • Height:
    5,5 mm
  • Length:
    25 mm