Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, rectangular

VWR®, Magnetic stirring bars, rectangular

12 mm, 4 mm
Catalog # 442-0486
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, rectangular
Catalog # 442-0486
Supplier:  VWR Collection
VWR®, Magnetic stirring bars, rectangular
VWR®, Magnetic stirring bars, rectangular

12 mm, 4 mm

Price
15,80 €
Log in for availability
1 * 10 ST
Product Details & Documents

PTFE covered, strong Alnico V magnetic core. Smooth surface. Very effective stirrers, especially at low speeds.

  • Excellent chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (−200 to +280 °C)
Specifications
  • Height:
    4 mm
  • Length:
    12 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.