VWR®, Magnetic stirring bars, triangular
12 mm, 6 mm
Catalog # 442-0385
Supplier: VWR Collection
Jump to:
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly effective for dissolving solids and mixing sediments because of the scraper-like action on the base of the container.
- High turbulence, even at low speeds
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
- Ø:6 mm
- Length:12 mm



