Orders LinkContactUsLinkComponent
Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
Catalog # KGWA1740
Supplier:   KGW
Cold traps, Type KF
Catalog # KGWA1740
Supplier:  KGW
Cold traps, Type KF
Cold traps, Type KF
Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
Price
579,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents. Model KF cold traps have a GL 32 glass thread together with a screw-on cap (spout), through which the condensate can easily be poured out of the cold trap. Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask made of blue coated metal or aluminium
Specifications
  • Description:
    Cold trap, KF 29-GL, borosilicate glass 3,3, with 150 ml condensate capacity and 1000 ml coolent cap
  • Seq:
    06
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Plastic ring for cold trap, type KF
    Plastic ring for cold trap, type KF
    KGWA1733
    Log in for availability
    1 * 1 ST - 62,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Cooling finger S 29-GL
    Cooling finger S 29-GL
    KGWA1741
    Log in for availability
    1 * 1 ST - 275,00 €