Orders LinkContactUsLinkComponent
Closures for test tubes and flasks
Closures for test tubes and flasks
Cotton balls, medium, non sterile
Catalog # 734-1266
Supplier:   US COTTON
Closures for test tubes and flasks
Catalog # 734-1266
Supplier:  US COTTON
Closures for test tubes and flasks
Closures for test tubes and flasks
Cotton balls, medium, non sterile
Price
44,81 €
Log in for availability
1 * 2.000 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.
Specifications
  • Description:
    Cotton balls, medium, non sterile
  • Seq:
    08