Order Entry
Belgium
Orders LinkContactUsLinkComponent
Conductivity standards

Conductivity standards

Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability, 147 µS/cm, 500 mL
Catalog # HAMI238985
Supplier:   HAMILTON BONADUZ
Conductivity standards
Catalog # HAMI238985
Supplier:  HAMILTON BONADUZ
CAS Number:  
Conductivity standards
Conductivity standards

Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability, 147 µS/cm, 500 mL

CAS Number:  
Price
67,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

Conductivity standards with stability of ±1% - guaranteed over a lifetime of up to three years.

  • Remains stable for a minimum of 1 year for 1,3 μs/cm, and up to 3 years for all other values
  • Certificate with calibration document from DFM
  • Expiration date shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes

The procedure for determining conductivity was developed in collaboration with DFM (Danish Institute of Fundamental metrology). A representative number of bottles from every batch are measured by DFM. Their value is recorded on the calibration certificate and on every bottle. DFM enjoys the highest prestige in Europe in the area of electrolytic conductivity and is equipped with an absolute measurement cell that was developed in collaboration with NIST, and is accredited by the Danish accreditation agency DANAK to a conductivity of 0,9 μs/cm.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Conductivity standard, Basic Line, in Calpack bottle, with 18 months stability
  • Conductivity:
    147 µS/cm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.