Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Cables for Electrodes

Plug cable combination, L 1 N (coaxial electrode plug, 1 m coaxial cable, 4 mm banana plug), 1 m
Catalog # 662-2192
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2192
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes

Plug cable combination, L 1 N (coaxial electrode plug, 1 m coaxial cable, 4 mm banana plug), 1 m

Price
48,40 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Plug cable combination, L 1 N (coaxial electrode plug, 1 m coaxial cable, 4 mm banana plug)
  • Length:
    1 m
  • Seq:
    21
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.