Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Fused and sealed at both ends, 6 mm, 200 mm
Catalog # 441-0146
Supplier:   Brand
Stirring rods
Catalog # 441-0146
Supplier:  Brand
Stirring rods
Stirring rods

Fused and sealed at both ends, 6 mm, 200 mm

Price
30,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

AR® glass, Glass tube is fused and sealed at both ends.

Specifications
  • Ø:
    6 mm
  • Description:
    Fused and sealed at both ends
  • Length:
    200 mm
  • Seq:
    03