Microscope slides, UniMark®
Slides in standard packs, Orange
N° de catalogue 631-1209
Fournisseur: MARIENFELD
Aller à :
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
- Frosted end for labelling
- Soda-lime glass, Hydrolytic Class 3
- Different colours to identify preparations securely
- Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Documents
Certificats
Rechercher par numéro de lotCoC_MARI1000612_823410
Déclaration de conformité
MARI0704102_TDS_EN
Fiche technique
Spécifications
- Colour:Orange
- Description:Slides in standard packs
- Seq:06