Orders LinkContactUsLinkComponent
Staining systems for microscope slides, DrainRack Junior
Staining systems for microscope slides, DrainRack Junior
Staining systems, White, 100
Catalog # 631-1918
Supplier:   Simport Scientific
Staining systems for microscope slides, DrainRack Junior
Catalog # 631-1918
Supplier:  Simport Scientific
Staining systems for microscope slides, DrainRack Junior
Staining systems for microscope slides, DrainRack Junior
Staining systems, White, 100
Price
55,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
This rugged tray used as a drain rack can hold up to 100 microscope slides in 50 individually numbered slots. All slides are stored upright for easier insertion and removal.
  • Barcodes can be read without having to remove the slides from the box
  • To save space, the number of slides in each slot can be doubled simply by inserting two slides in each slot
  • Not autoclavable
Specifications
  • No. of slides:
    100
  • Colour:
    White
  • Description:
    Staining systems