Orders LinkContactUsLinkComponent
Face shields, Bionic™
Face shields, Bionic™
Uncoated, PC
Catalog # 111-0133
Supplier:   HONEYWELL SAFETY
Face shields, Bionic™
Catalog # 111-0133
Supplier:  HONEYWELL SAFETY
Face shields, Bionic™
Face shields, Bionic™
Uncoated, PC
Price
51,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
This innovative face shield provides protection from extreme mechanical hazards. It can be worn in combination with safety spectacles or respiratory protection.
  • With extended protection for brow and chin
  • Locking mechanism for added safety
  • Extra soft padding with washable or replaceable sweatband
  • Various adjustments for best comfort
Weight: 355 g.
EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Certifications: EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
Specifications
  • Type:
    Uncoated
  • Material:
    PC
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, uncoated
    Replacement visor, uncoated
    111-0740
    PC
    Log in for availability
    1 * 1 ST - 12,60 €
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, uncoated
    Replacement visor, uncoated
    111-2160
    Acetate
    Log in for availability
    1 * 1 ST - 20,60 €
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement visor, anti-scratch/anti-fog
    Replacement visor, anti-scratch/anti-fog
    111-1293
    PC
    Log in for availability
    1 * 1 ST - 32,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement sweatband
    Replacement sweatband
    113-8053
    Log in for availability
    1 * 1 ST - 7,40 €