Goggles, Flex Seal™
Anti-UV, anti-scratch, anti-fog, Clear PC, Transparent
Catalog # 111-0132
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
Goggles with a flexible and sturdy frame feature a wraparound lens for a panoramic view.
- Soft and flexible silicone frame for excellent comfort and ensures a snug, slip-free fit
- Remains soft and supple even in extremely hot or cold environments
- Pivoting, removable and adjustable headband
- Indirect ventilation prevents fogging
- Neoprene strap (111-0132) or elastic strap (111-3606)
Designed to be compatible with prescription spectacles.
Weight: 110 g.
: EN 166, EN 169, EN 171, 1FT, C 3, 4, 9.
Specifications
- Type:Anti-UV, anti-scratch, anti-fog
- Colour:Transparent
- Lenses:Clear PC