Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions

pH buffer solutions

Technical buffer solution, 6×250 ml, 4,64, Colourless, 250 ml, 6
Catalog # METN51319014
pH buffer solutions
Catalog # METN51319014
Supplier:  
CAS Number:  
pH buffer solutions
pH buffer solutions

Technical buffer solution, 6×250 ml, 4,64, Colourless, 250 ml, 6

CAS Number:  
Discontinued
Product Details & Documents
Specifications

Specifications

  • Pk:
    6
  • Colour:
    Colourless
  • Description:
    Technical buffer solution, 6×250 ml
  • Pk Info:
    250 ml
  • pH value:
    4,64

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.