Orders LinkContactUsLinkComponent
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, parallel, 4,0 mm, 18,8 NS
Catalog # 189-2112
Supplier:   Lenz Laborglas
Vacuum stopcocks
Catalog # 189-2112
Supplier:  Lenz Laborglas
Vacuum stopcocks
Vacuum stopcocks
Vacuum stopcocks, one way, parallel, 4,0 mm, 18,8 NS
Price
71,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
Specifications
  • Hole Ø:
    4,0 mm
  • Description:
    Vacuum stopcocks, one way, parallel
  • Seq:
    02
  • Spigot:
    18,8 NS