Volumetric flasks
GL 18
Catalogus # 612-4905
Leverancier: Brand
Ga naar:
Product Details & Documents
PFA, translucent, with PFA screw cap.
- Inclusive one batch certificate
- Exposure to temperatures up to 121 ºC (autoclaving) will not cause permanently exceeded error limits
- Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
Error limits according to class A, DIN EN ISO 1042. Calibrated to contain (TC, In). PFA screw cap with molded sealing ring. Autoclavable, easy to clean.
*Wide neck.
(Height = Height without stopper).
Documents
Brand PFA Labware Brand Volumetric Measurement in the Laboratory
Product documentatie
BRAND Volumetric Measurement
Video 's
TDS PFA labware.pdf TDS volumetric measurement
Technische fiche
Specificaties
- Thread:GL 18