Vortex mixer, VORTEX Genius 3
VORTEX Genius 3, EU-plug
Catalog # 412-0098
Supplier: IKA
Jump to:
Product Details & Documents
Vortex mixer with three interchangeable attachments and various inserts for different applications. Microcentrifuge tubes, microtitre plates, even round bottomed or 250 ml Erlenmeyer flasks can be attached by means of the specially designed strap. Attachments click securely onto the unit in any position. The unit is stable at high speeds and sturdy with coated, zinc die-casting.
- Compact design
- Wide speed range, infinitely adjustable
- Touch or continuous mode
- Can be used at 5 to 40 °C (80% relative humidity) in an incubator or cold room
IP protection class according to DIN EN 60529: IP 21
* Depending on attachment and loading
Certifications: IP protection class according to DIN EN 60529: IP 21
Ordering information: Supplied with standard attachment for test tubes or small beakers, 412-0099.
Caution: * Depending on attachment and loading
** Use in combination with 412-0101
Specifications
- Description:VORTEX Genius 3, EU-plug
- Shaking motion:Orbital
- Cat. No.:412-0098
- Motor input/output:58/10 W
- Orbit:4 mm
- Speed range:500 - 2500 min⁻¹*
- Model:VORTEX Genius 3
- W×D×H:127×149×136 mm
- Weight:4,5 kg
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Flask attachment for Erlenmeyer or round bottom flasks, 100 up to 250 ml**Flask attachment for Erlenmeyer or round bottom flasks, 100 up to 250 ml**412-0107Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 8 reagent tubes, 16 mm**Test tube attachment, for 8 reagent tubes, 16 mm**412-0105Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 18 reagent tubes, 10 mm**Test tube attachment, for 18 reagent tubes, 10 mm**412-0103Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 12 reagent tubes, 12 mm**Test tube attachment, for 12 reagent tubes, 12 mm**412-0104Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 54 microcentrifuge tubes, 1,5/2,0 ml**Test tube attachment, for 54 microcentrifuge tubes, 1,5/2,0 ml**412-0102Continuous modeLog in for availability1 * 1 ST - 81,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Test tube attachment, for 8 reagent tubes, 20 mm**Test tube attachment, for 8 reagent tubes, 20 mm**412-0106Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Microtitre plate attachment for 1 standard microtitre plate**Microtitre plate attachment for 1 standard microtitre plate**441-0036Continuous modeLog in for availability1 * 1 ST - 43,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Attachment for single-handed operation, 88 mm round with rubber insertAttachment for single-handed operation, 88 mm round with rubber insert412-0100Continuous or touch modeLog in for availability1 * 1 ST - 31,50 €
Product Information
- Catalog #ForAvailabilityPrice
- Universal attachment, 150 mm with rubber insertUniversal attachment, 150 mm with rubber insert412-0101Continuous modeLog in for availability1 * 1 ST - 35,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Standard attachment



