Orders LinkContactUsLinkComponent
Watchmaker forceps
Watchmaker forceps
Watchmaker No. 4, straight, 110 mm
N° de catalogue 232-0202
Fournisseur:   MURRAY S
Watchmaker forceps
N° de catalogue 232-0202
Fournisseur:  MURRAY S
Watchmaker forceps
Watchmaker forceps
Watchmaker No. 4, straight, 110 mm
Prix
12,10 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Stainless steel. Ideal for small-scale dissection applications or precise part placement.
  • Very fine points
  • Smooth flat handle
  • Available straight or curved
Spécifications
  • Type:
    Watchmaker No. 4, straight
  • Length:
    110 mm