Order Entry
Belgium
Orders LinkContactUsLinkComponent
Spray bottles

Spray bottles

500 ml, 206 mm
Catalog # MURRP01422-P51314
Supplier:   MURRAY S
Spray bottles
Catalog # MURRP01422-P51314
Supplier:  MURRAY S
Spray bottles
Spray bottles

500 ml, 206 mm

Price
471,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

HDPE, translucent.

Designed to allow comfortable grip and minimise fatigue. Adjustable from fine spray to single jet. Delivery 1,4 ml per stroke.

Specifications
  • Capacity:
    500 ml
  • Height:
    206 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.