Order Entry
Belgium
Orders LinkContactUsLinkComponent
Safe-Lock Tubes, 1,5 ml - Microtubes

Safe-Lock Tubes, 1,5 ml - Microtubes

1,5 ml, Colourless, Safe-Lock, PP, 22000×g, 100 tubes, individually wrapped
Catalog # 211-2161
   Low Manufacturing Impact
Supplier:   EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Catalog # 211-2161
Supplier:  EPPENDORF
   Low Manufacturing Impact
Safe-Lock Tubes, 1,5 ml - Microtubes
Safe-Lock Tubes, 1,5 ml - Microtubes

1,5 ml, Colourless, Safe-Lock, PP, 22000×g, 100 tubes, individually wrapped

Price
36,40 €
Log in for availability
1 * 100 ST
Product Details & Documents

Eppendorf® Safe-Lock 1,5 ml microtubes ensure secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1,5 ml capacity.

  • Hinged Safe-Lock lid prevents inadvertent lid opening during incubation and storage
  • g-Safe® technology, depending on the Safe-Lock Tube variant, ensures outstanding centrifugation stability up to 30,000×g
  • g-Safe® technology prevents sample loss due to tube breakage, and provides additional safety when handling hazardous samples
  • Polypropylene construction offers robust resistance to chemicals, mechanical stress, and extreme temperatures
  • Precision lid sealing minimises evaporation rates during extended storage periods
  • Generously sized frosted lid and side surface facilitate easy labelling
  • Assured functionality in a temperature range from −86 to 100 °C
  • Autoclavable when open (121 °C, 20 min) for reliable sterilisation
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA Grade, and Biopur® quality for tested purity

Eppendorf® Safe-Lock 1,5 ml microtubes exemplify secure sample storage with their patented Safe-Lock design, preventing accidental openings and preserving sample integrity. Versatile across laboratory applications, the durable construction and 1,5 ml capacity cater to varied sample volumes.

Clear graduations and a smooth surface enhance user-friendliness, ensuring a seamless and reliable laboratory experience. Elevate scientific endeavors with the trusted quality of Eppendorf® Safe-Lock Tubes.

Manufactured without the use of slip agents, plasticisers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA Grade can be downloaded from the Eppendorf website.

Specifications
  • Packed:
    100 tubes, individually wrapped
  • Type:
    Safe-Lock
  • Capacity:
    1,5 ml
  • Colour:
    Colourless
  • Material:
    PP
  • Seq:
    09
  • RCF max.:
    22000×g
  • Cat. No.:
    211-2161
  • Operating temperature range:
    −86...100 °C
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Volume:
    1,5 ml
  • Autoclavable:
    When open 121 °C, 20 min
  • Environmentally Preferable:
    Low Manufacturing Impact