Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, Combined pH Electrodes, pHASE

VWR®, Combined pH Electrodes, pHASE

Sealed, flat tip, double junction (HDPE), 0 - 14, Epoxy, Gel, 12×150 mm, BNC, 150 mm, Aqueous media, beverage, Tris buffer, cosmetics, ultra-pure water
Catalog # 662-1780
Supplier:   VWR Collection
VWR®, Combined pH Electrodes, pHASE
Catalog # 662-1780
Supplier:  VWR Collection
VWR®, Combined pH Electrodes, pHASE
VWR®, Combined pH Electrodes, pHASE

Sealed, flat tip, double junction (HDPE), 0 - 14, Epoxy, Gel, 12×150 mm, BNC, 150 mm, Aqueous media, beverage, Tris buffer, cosmetics, ultra-pure water

Price
337,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Combined electrodes providing fast, accurate and stable pH measurements, ideal for control quality and research and development use.

  • Fast, accurate and stable pH measurements, fast thermal equilibration, high accuracy even in samples with large temperature range
  • Available with sealed design (economical, easy to use, with minimum maintenance) or Rapid-Renew design (refillable electrodes used with dirty or other samples that cause junction fouling)
  • Available with epoxy body (with good chemical resistance, maximum ruggedness and removable bulb guard) or glass body (maximum chemical resistance with fully exposed ruggedised bulbs)
  • With 1 m fixed cable

pH range: 0 to 14

Accuracy: to 0,04 pH between 0 to 100 °C in pH 7 buffer

Stability: 0,005 pH or less drift per hour

Temperature range: 0 to +100 °C

Temperature range: 0 to +100 °C

Specifications
  • Connection:
    BNC
  • Ø×L:
    12×150 mm
  • For:
    Aqueous media, beverage, Tris buffer, cosmetics, ultra-pure water
  • Type:
    Sealed, flat tip, double junction (HDPE)
  • Length:
    150 mm
  • Material:
    Epoxy
  • Seq:
    06
  • Electrolyte:
    Gel
  • pH:
    0 - 14
Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Refill solution 3,5 mol/l KCl
    Refill solution 3,5 mol/l KCl
    662-1784
    100 mL
    Log in for availability
    1 * 100 mL - 56,50 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.